Merck
PKA-RIIβ autophosphorylation modulates PKA activity and seizure phenotypes in mice.
Communications biology (2021-03-03)
Jingliang Zhang, Chenyu Zhang, Xiaoling Chen, Bingwei Wang, Weining Ma, Yang Yang, Ruimao Zheng, Zhuo Huang
ZUSAMMENFASSUNG
Temporal lobe epilepsy (TLE) is one of the most common and intractable neurological disorders in adults. Dysfunctional PKA signaling is causally linked to the TLE. However, the mechanism underlying PKA involves in epileptogenesis is still poorly understood. In the present study, we found the autophosphorylation level at serine 114 site (serine 112 site in mice) of PKA-RIIβ subunit was robustly decreased in the epileptic foci obtained from both surgical specimens of TLE patients and seizure model mice. The p-RIIβ level was negatively correlated with the activities of PKA. Notably, by using a P-site mutant that cannot be autophosphorylated and thus results in the released catalytic subunit to exert persistent phosphorylation, an increase in PKA activities through transduction with AAV-RIIβ-S112A in hippocampal DG granule cells decreased mIPSC frequency but not mEPSC, enhanced neuronal intrinsic excitability and seizure susceptibility. In contrast, a reduction of PKA activities by RIIβ knockout led to an increased mIPSC frequency, a reduction in neuronal excitability, and mice less prone to experimental seizure onset. Collectively, our data demonstrated that the autophosphorylation of RIIβ subunit plays a critical role in controlling neuronal and network excitabilities by regulating the activities of PKA, providing a potential therapeutic target for TLE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-GAPDH-Antikörper, Maus monoklonal in Maus hergestellte Antikörper, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution