Merck
Pharmacological characterization of SIB-1765F: a novel cholinergic ion channel agonist.
The Journal of pharmacology and experimental therapeutics (1997-01-01)
A I Sacaan, R T Reid, E M Santori, P Adams, L D Correa, L S Mahaffy, L Bleicher, N D Cosford, K A Stauderman, I A McDonald, T S Rao, G K Lloyd
ZUSAMMENFASSUNG
Nicotine, the prototypical agonist for neuronal nicotinic acetylcholine receptors (NAChR), nonselectively activates NAChR limiting its use in elucidating the function of NAChR subtypes. SIB-1765F is a subtype selective NAChR agonist that displaces [3H]-nicotine binding with an IC50 of 4.6 nM and [3H]-cytisine binding with an IC50 of 12.2 nM which is 2000- to 6000-fold lower than its displacement of [3H]-QNB or [125I]-alpha-bungarotoxin. SIB-1765F did not inhibit human or rat cholinesterases or the uptake of [3H]-DA in synaptosomal preparations. SIB-1765F mimicked (-)-nicotine in stimulating [3H]-DA release from rat striatal and olfactory tubercle slices, with EC50 values of 99.6 and 39.6 microM, respectively. Such stimulation was sensitive to mecamylamine and DH beta E. SIB-1765F also released endogenous DA in the striatum and the nucleus accumbens as measured by in vivo microdialysis. SIB-1765F was less efficacious than (-)-nicotine at stimulating [3H]-NE release from rat hippocampal slices; in contrast, SIB-1765F increased [3H]-NE release from rat thalamic and cortical slices with efficacies approaching those of (-)-nicotine. Similar to (-)-nicotine and (+/-)-epibatidine, subcutaneous administration of SIB-1765F increased the turnover rate of dopamine ex vivo both in the striatum and olfactory tubercles in a mecamylamine-sensitive manner. Because the release of striatal DA and hippocampal NE appears to be regulated by distinct NAChR, differential effects of SIB-1765F on striatal DA and hippocampal NE release supports the NAChR subtype selectivity of SIB-1765F compared to (-)-nicotine. This is further demonstrated by observations showing that SIB-1765F has a higher affinity for h alpha 4 beta 2 NAChR relative to h alpha 4 beta 4 NAChRs in displacing [3H]-epibatidine binding and increasing cytosolic CA+2 concentration in cell lines stably expressing h alpha 4 beta 2 or h alpha 4 beta 4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Butyrylcholinesterase aus Humanserum, vial of ≥4 units