Merck
HDAC1 SUMOylation promotes Argonaute-directed transcriptional silencing in C. elegans.
eLife (2021-05-19)
Heesun Kim, Yue-He Ding, Gangming Zhang, Yong-Hong Yan, Darryl Conte, Meng-Qiu Dong, Craig C Mello
ZUSAMMENFASSUNG
Eukaryotic cells use guided search to coordinately control dispersed genetic elements. Argonaute proteins and their small RNA cofactors engage nascent RNAs and chromatin-associated proteins to direct transcriptional silencing. The small ubiquitin-like modifier (SUMO) has been shown to promote the formation and maintenance of silent chromatin (called heterochromatin) in yeast, plants, and animals. Here, we show that Argonaute-directed transcriptional silencing in Caenorhabditis elegans requires SUMOylation of the type 1 histone deacetylase HDA-1. Our findings suggest how SUMOylation promotes the association of HDAC1 with chromatin remodeling factors and with a nuclear Argonaute to initiate de novo heterochromatin silencing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
TRI-Reagenz®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Ampicillin Natriumsalz
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
1-Brom-3-chlorpropan, for isolation of RNA
Sigma-Aldrich
Calciumchlorid Dihydrat, BioXtra, ≥99.0%
Sigma-Aldrich
Imidazol, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Anti-trimethyl-Histon H3-(Lys4-)Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%
Roche
Lumi-LightPLUS Western-Blot-Substrat, pkg of 100 mL (1,000 cm2 membrane)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, serum, Upstate®