Merck
Detection of pathological alpha-synuclein aggregates in human iPSC-derived neurons and tissue.
STAR protocols (2021-03-19)
Iva Stojkovska, Joseph R Mazzulli
ZUSAMMENFASSUNG
The accumulation of proteins into insoluble aggregates is a common feature of several neurodegenerative diseases. Aggregated α-synuclein is a major component of Lewy bodies that pathologically define Parkinson's disease (PD). Here, we present methods for the detection of pathogenic conformations of α-synuclein in induced pluripotent stem cell (iPSC) patient-derived neuron models and brain tissue. These methods can be applied to studies of PD pathogenesis and the discovery of novel therapeutics that restore physiological α-synuclein. For complete details on the use and execution of this protocol, please refer to Cuddy et al. (2019) and Zunke et al. (2018).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Magnesiumchlorid Hexahydrat, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Thioflavin S, practical grade
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%