Merck
  • Variability in Zinc Concentration among Mueller-Hinton Broth Brands: Impact on Antimicrobial Susceptibility Testing of Metallo-β-Lactamase-Producing Enterobacteriaceae.
Variability in Zinc Concentration among Mueller-Hinton Broth Brands: Impact on Antimicrobial Susceptibility Testing of Metallo-β-Lactamase-Producing Enterobacteriaceae.
Journal of clinical microbiology (2020-10-02)
Anastasia Bilinskaya, Douglas J Buckheit, Michael Gnoinski, Tomefa E Asempa, David P Nicolau
ZUSAMMENFASSUNG
Zinc concentrations in cation-adjusted Mueller-Hinton broth (caMHB) from different manufacturers have been found to differ. Here, we evaluated the impact of utilizing different brands and lots of commercially available caMHB on the classification of the antimicrobial susceptibility of metallo-β-lactamase (MBL)-harboring Enterobacteriaceae We also evaluated the addition of EDTA to caMHB as a means of achieving zinc-limited media. Fifteen clinical Enterobacteriaceae isolates (harboring NDM [n = 7], VIM [n = 3], IMP [n = 2], or KPC [n = 3]) and nine different commercial lots from three caMHB manufacturers (Becton, Dickinson; Oxoid; and Sigma-Aldrich) were utilized. Zinc-limited media were prepared by the addition of EDTA at concentrations ranging from 3 to 300 μg/ml. Meropenem MICs were determined in triplicate for each lot of conventional caMHB and zinc-limited media by broth microdilution. The zinc concentration in each lot of conventional caMHB was determined by inductively coupled plasma mass spectrometry. Up to 8-fold differences in meropenem MICs were observed between the commercial lots, resulting in different classifications of susceptibility among MBL-harboring isolates. Mean zinc concentrations were highest among conventional Becton, Dickinson caMHB lots relative to those for Oxoid and Sigma-Aldrich broth. Among MBL-harboring isolates, the impact of EDTA on MICs was dependent on the lot, correlating with initial zinc availability (i.e., less MIC reduction with higher initial zinc concentrations), while MICs for KPC-harboring isolates were unchanged. In summary, zinc variability was observed among commercial lots of caMHB, resulting in different classifications of susceptibility among MBL-harboring Enterobacteriaceae The addition of EDTA at concentrations of ≥30 μg/ml was sufficient to provide a zinc-limited medium, resulting in MICs that reflect in vivo meropenem activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
EDTA, Dinatriumsalz, Dihydrat, molekularbiologische Qualität