- Infectious Subviral Particle-induced Hemolysis Assay for Mammalian Orthoreovirus.
Infectious Subviral Particle-induced Hemolysis Assay for Mammalian Orthoreovirus.
Bio-protocol (2018-03-20)
Anthony J Snyder, Pranav Danthi
PMID29552594
ZUSAMMENFASSUNG
Mammalian orthoreovirus (reovirus) utilizes pore forming peptides to penetrate host cell membranes. This step is essential for delivering its genome containing core particle during viral entry. This protocol describes an in vitro assay for measuring reovirus-induced pore formation.