Merck
  • A protocol for simultaneous Ca2+ and morphology imaging of brain endothelial tip cells in larval zebrafish.
A protocol for simultaneous Ca2+ and morphology imaging of brain endothelial tip cells in larval zebrafish.
STAR protocols (2021-03-30)
Ting-Ting Liu, Han Hou, Jiu-Lin Du
ZUSAMMENFASSUNG
Endothelial tip cells (ETCs) located at growing blood vessels display high morphological dynamics and associated intracellular Ca2+ activities with different spatiotemporal patterns during migration. Examining the Ca2+ activity and morphological dynamics of ETCs will provide an insight for understanding the mechanism of vascular development in organs, including the brain. Here, we describe a method for simultaneous monitoring and relevant analysis of the Ca2+ activity and morphology of growing brain ETCs in larval zebrafish. For complete details on the use and execution of this protocol, please refer to Liu et al. (2020).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesiumchlorid Hexahydrat, ACS reagent, 99.0-102.0%