Merck
Comparison of titanium soaked in 5 M NaOH or 5 M KOH solutions.
Materials science & engineering. C, Materials for biological applications (2013-04-09)
Christina Kim, Matthew R Kendall, Matthew A Miller, Courtney L Long, Preston R Larson, Mary Beth Humphrey, Andrew S Madden, A Cuneyt Tas
ZUSAMMENFASSUNG
Commercially pure titanium plates/coupons and pure titanium powders were soaked for 24 h in 5 M NaOH and 5 M KOH solutions, under identical conditions, over the temperature range of 37° to 90 °C. Wettability of the surfaces of alkali-treated cpTi coupons was studied by using contact angle goniometry. cpTi coupons soaked in 5 M NaOH or 5 M KOH solutions were found to have hydrophilic surfaces. Hydrous alkali titanate nanofibers and nanotubes were identified with SEM/EDXS and grazing incidence XRD. Surface areas of Ti powders increased > 50–220 times, depending on the treatment, when soaked in the above solutions. A solution was developed to coat amorphous calcium phosphate, instead of hydroxyapatite, on Ti coupon surfaces. In vitro cell culture tests were performed with osteoblast-like cells on the alkali-treated samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%