Merck
  • Features of the Synthesis of the Dispersed TiC Phase with Nickel Nanostructures on the Surface to Create an Aluminum-Based Metal Composite.
Features of the Synthesis of the Dispersed TiC Phase with Nickel Nanostructures on the Surface to Create an Aluminum-Based Metal Composite.
Nanomaterials (Basel, Switzerland) (2021-10-24)
Elena G Zemtsova, Denis V Yurchuk, Pavel E Morozov, Petr M Korusenko, Vladimir K Kudymov, Vladimir M Smirnov
ZUSAMMENFASSUNG
The development of new composites with improved functional properties is the important task of modern materials science. The composites must be structurally organized to provide improved properties. For metal-ceramic composites, there is a need for a uniform distribution of the dispersed ceramic phase in the bulk metallic matrix The modification of the dispersed ceramic phase surface with a metal coating is one of the more effective ways to accomplish this. Particularly, in this work, the conditions of Ni nanolayer deposition on titanium carbide (TiC) particles were studied. The goal was to create core-shell particles with a thickness of the Ni coating on TiC not exceeding 90 nm. Preliminary work was also carried out to study the effect of the dispersed phase composition on the mechanical properties of the composite with an Al matrix.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Natriumcitrat-Dihydrat, ≥99%, FG