Merck
  • αE-catenin actin-binding domain alters actin filament conformation and regulates binding of nucleation and disassembly factors.
αE-catenin actin-binding domain alters actin filament conformation and regulates binding of nucleation and disassembly factors.
Molecular biology of the cell (2013-09-27)
Scott D Hansen, Adam V Kwiatkowski, Chung-Yueh Ouyang, Hongjun Liu, Sabine Pokutta, Simon C Watkins, Niels Volkmann, Dorit Hanein, William I Weis, R Dyche Mullins, W James Nelson
ZUSAMMENFASSUNG
The actin-binding protein αE-catenin may contribute to transitions between cell migration and cell-cell adhesion that depend on remodeling the actin cytoskeleton, but the underlying mechanisms are unknown. We show that the αE-catenin actin-binding domain (ABD) binds cooperatively to individual actin filaments and that binding is accompanied by a conformational change in the actin protomer that affects filament structure. αE-catenin ABD binding limits barbed-end growth, especially in actin filament bundles. αE-catenin ABD inhibits actin filament branching by the Arp2/3 complex and severing by cofilin, both of which contact regions of the actin protomer that are structurally altered by αE-catenin ABD binding. In epithelial cells, there is little correlation between the distribution of αE-catenin and the Arp2/3 complex at developing cell-cell contacts. Our results indicate that αE-catenin binding to filamentous actin favors assembly of unbranched filament bundles that are protected from severing over more dynamic, branched filament arrays.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP