Merck
Encephalopathy-causing mutations in Gβ1 (GNB1) alter regulation of neuronal GIRK channels.
iScience (2021-09-16)
Haritha P Reddy, Daniel Yakubovich, Tal Keren-Raifman, Galit Tabak, Vladimir A Tsemakhovich, Maria H Pedersen, Boris Shalomov, Sophie Colombo, David B Goldstein, Jonathan A Javitch, Amal K Bera, Nathan Dascal
ZUSAMMENFASSUNG
Mutations in the GNB1 gene, encoding the Gβ1 subunit of heterotrimeric G proteins, cause GNB1 Encephalopathy. Patients experience seizures, pointing to abnormal activity of ion channels or neurotransmitter receptors. We studied three Gβ1 mutations (K78R, I80N and I80T) using computational and functional approaches. In heterologous expression models, these mutations did not alter the coupling between G protein-coupled receptors to Gi/o, or the Gβγ regulation of the neuronal voltage-gated Ca2+ channel CaV2.2. However, the mutations profoundly affected the Gβγ regulation of the G protein-gated inwardly rectifying potassium channels (GIRK, or Kir3). Changes were observed in Gβ1 protein expression levels, Gβγ binding to cytosolic segments of GIRK subunits, and in Gβγ function, and included gain-of-function for K78R or loss-of-function for I80T/N, which were GIRK subunit-specific. Our findings offer new insights into subunit-dependent gating of GIRKs by Gβγ, and indicate diverse etiology of GNB1 Encephalopathy cases, bearing a potential for personalized treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Calciumchlorid, anhydrous, granular, ≤7.0 mm, ≥93.0%
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Ampicillin Natriumsalz
Sigma-Aldrich
Poly-L-Lysin -hydrobromid, mol wt 30,000-70,000
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%