Merck
Genetically Engineered Mouse Model of Brainstem High-Grade Glioma.
STAR protocols (2020-12-31)
Fernando M Nunez, Jessica C Gauss, Flor M Mendez, Santiago Haase, Pedro R Lowenstein, Maria G Castro
ZUSAMMENFASSUNG
Brainstem gliomas are aggressive tumors that are more prevalent in pediatric patients. The location of these tumors makes them inoperable, and currently there is no effective treatment. Recent genomic data revealed the unique biology of these tumors. The following protocol provides a method to incorporate these specific genetic lesions in a mouse glioma model. Using this model, the effects of these mutations in tumor progression and response to treatments can be studied within a relevant in vivo context. For complete details on the use and execution of this protocol, please refer to Mendez et al. (2020).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Tyrode′s Salts, Without sodium bicarbonate, powder, suitable for cell culture