- A protocol for in vivo analysis of liver tumorigenesis in mice using sleeping beauty transposon system.
A protocol for in vivo analysis of liver tumorigenesis in mice using sleeping beauty transposon system.
STAR protocols (2021-04-30)
Lingli He, Wentao Yu, Yi Lu, Wenxiang Zhang, Jinjin Xu, Lei Zhang
PMID33912846
ZUSAMMENFASSUNG
Hepatocellular carcinoma (HCC) is the fourth leading cause of cancer death worldwide. However, the pathogenesis of HCC is complicated, and the drugs used for HCC treatment are limited. The following protocol combines a genetically engineered mouse model (GEMM) with a sleeping beauty system to establish an in vivo liver tumorigenesis model. By using this model, the impact of genes of interest in liver tumorigenesis and progression can be studied. This model can also be applied to develop new therapeutic drugs for HCC. For complete details on the use and execution of this protocol, please refer to He et al. (2020).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)