Merck
Bacterial Factors Required for Transmission of Streptococcus pneumoniae in Mammalian Hosts.
Cell host & microbe (2019-05-28)
Hannah M Rowe, Erik Karlsson, Haley Echlin, Ti-Cheng Chang, Lei Wang, Tim van Opijnen, Stanley B Pounds, Stacey Schultz-Cherry, Jason W Rosch
ZUSAMMENFASSUNG
The capacity of Streptococcus pneumoniae to successfully transmit and colonize new human hosts is a critical aspect of pneumococcal population biology and a prerequisite for invasive disease. However, the bacterial mechanisms underlying this process remain largely unknown. To identify bacterial factors required for transmission, we conducted a high-throughput genetic screen with a transposon sequencing (Tn-seq) library of a pneumococcal strain in a ferret transmission model. Key players in both metabolism and transcriptional regulation were identified as required for efficient bacterial transmission. Targeted deletion of the putative C3-degrading protease CppA, iron transporter PiaA, or competence regulatory histidine kinase ComD significantly decreased transmissibility in a mouse model, further validating the screen. Maternal vaccination with recombinant surface-exposed PiaA and CppA alone or in combination blocked transmission in offspring and were more effective than capsule-based vaccines. These data underscore the possibility of targeting pneumococcal transmission as a means of eliminating invasive disease in the population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kohlensäure-Bicarbonat-Puffer, capsule