Merck
  • ABPP-HT - High-Throughput Activity-Based Profiling of Deubiquitylating Enzyme Inhibitors in a Cellular Context.
ABPP-HT - High-Throughput Activity-Based Profiling of Deubiquitylating Enzyme Inhibitors in a Cellular Context.
Frontiers in chemistry (2021-03-16)
Hannah B L Jones, Raphael Heilig, Roman Fischer, Benedikt M Kessler, Adán Pinto-Fernández
ZUSAMMENFASSUNG
The potency and selectivity of a small molecule inhibitor are key parameters to assess during the early stages of drug discovery. In particular, it is very informative for characterizing compounds in a relevant cellular context in order to reveal potential off-target effects and drug efficacy. Activity-based probes are valuable tools for that purpose, however, obtaining cellular target engagement data in a high-throughput format has been particularly challenging. Here, we describe a new methodology named ABPP-HT (high-throughput-compatible activity-based protein profiling), implementing a semi-automated proteomic sample preparation workflow that increases the throughput capabilities of the classical ABPP workflow approximately ten times while preserving its enzyme profiling characteristics. Using a panel of deubiquitylating enzyme (DUB) inhibitors, we demonstrate the feasibility of ABPP-HT to provide compound selectivity profiles of endogenous DUBs in a cellular context at a fraction of time as compared to previous methodologies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Triethylammoniumbicarbonat-Puffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
Magnesiumchlorid Hexahydrat, BioXtra, ≥99.0%
Sigma-Aldrich
DUB-Inhibitor V, PR-619, Calbiochem, The DUB Inhibitor V, PR-619 controls the biological activity of DUB. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
HBX 41,108, ≥98% (HPLC)