Merck
  • Mutagenesis of the active site lysine 221 of the pyruvate kinase from Bacillus stearothermophilus
Mutagenesis of the active site lysine 221 of the pyruvate kinase from <I>Bacillus stearothermophilus</I>
The Journal of Biological Chemistry (2005)
Sakai, H.