Merck
  • Zirconium (IV) Chloride Mediated Cyclodehydration of 1, 2-Diacylhydrazines: A Convenient Synthesis of 2, 5-Diaryl 1, 3, 4-Oxadiazoles
Zirconium (IV) Chloride Mediated Cyclodehydration of 1, 2-Diacylhydrazines: A Convenient Synthesis of 2, 5-Diaryl 1, 3, 4-Oxadiazoles
Synthetic Communications (2004)
Sharma GVM, et al.