Merck
  • Investigation of the Influence of Protein-Losing Enteropathy on Monoclonal Antibody Pharmacokinetics in Mice.
Investigation of the Influence of Protein-Losing Enteropathy on Monoclonal Antibody Pharmacokinetics in Mice.
The AAPS journal (2017-08-30)
Yujie Yang, Tommy R Li, Joseph P Balthasar
ZUSAMMENFASSUNG
Protein losing enteropathy (PLE), which is characterized by substantial loss of plasma proteins into the gastrointestinal (GI) tract, is a complication of a variety of GI diseases, including inflammatory bowel disease. Clinical studies have found that the clearance of monoclonal antibodies (mAb) is often increased in subjects with diseases known to cause PLE; however, direct relationships between PLE and mAb pharmacokinetics have not been demonstrated. This study employed a murine model of colitis to examine the influence of PLE on mAb pharmacokinetics. Mice were given dextran sodium sulfate (DSS, 2% w/v) supplemented tap water as drinking source for 6 days to induce colitis and PLE. Mice were then intravenously injected with 8C2, a murine IgG1 mAb. 8C2 plasma concentrations were measured up to 14 days post injection. Fecal alpha-1-antitrypsin (A1AT) clearance was measured as biomarker for PLE. DSS-treated mice developed PLE of clinically relevant severity. They also showed a transient increase in 8C2 plasma clearance and a decrease in 8C2 plasma exposure. The area under the 8C2 plasma concentration-time curve for the length of the study (AUC0-14d) reduced from 1368 ± 255 to 594 ± 224 day μg/ml following DSS treatment (p = 0.001). A quantitative relationship between A1AT clearance and 8C2 clearance was obtained via population pharmacokinetic modeling. DSS treatment substantially increased 8C2 clearance and reduced 8C2 exposure. Increased mAb plasma clearance was highly correlated with A1AT fecal clearance, suggesting the possible utility of A1AT fecal clearance as a mechanistic biomarker to predict the pharmacokinetics of therapeutic antibodies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)−Alkalische Phosphatase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution