Merck
  • Microbial production of 3-hydroxydodecanoic acid by pha operon and fadBA knockout mutant of Pseudomonas putida KT2442 harboring tesB gene.
Microbial production of 3-hydroxydodecanoic acid by pha operon and fadBA knockout mutant of Pseudomonas putida KT2442 harboring tesB gene.
Applied microbiology and biotechnology (2009-03-10)
Ahleum Chung, Qian Liu, Shao-Ping Ouyang, Qiong Wu, Guo-Qiang Chen
ZUSAMMENFASSUNG
To produce extracellular chiral 3-hydroxyacyl acids (3HA) by fermentation, a novel pathway was constructed by expressing tesB gene encoding thioesterase II into Pseudomonas putida KTOY01, which was a polyhydroxyalkanoate (PHA) synthesis operon knockout mutant. 3HA mixtures of 0.35 g/l consisting of 3-hydroxyhexanoate, 3-hydroxyoctanoate, 3-hydroxydecanoate, and 3-hydroxydodecanoate (3HDD) were produced in shake-flask study using dodecanoate as a sole carbon source. Additional knockout of fadB and fadA genes encoding 3-ketoacyl-CoA thiolase and 3-hydroxyacyl-CoA dehydrogenase in P. putida KTOY01 led to the weakening of the beta-oxidation pathway. The fadBA and PHA synthesis operon knockout mutant P. putida KTOY07 expressing tesB gene produced 2.44 g/l 3HA, significantly more than that of the beta-oxidation intact mutant. The 3HA mixture contained 90 mol% 3HDD as a dominant component. A fed-batch fermentation process carried out in a 6-l automatic fermentor produced 7.27 g/l extracellular 3HA containing 96 mol% fraction of 3HDD after 28 h of growth. For the first time, it became possible to produce 3HDD-dominant 3HA monomers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-β-Hydroxy-laurinsäure, ≥99% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.