Merck
Identification of a class of non-conventional ER-stress-response-derived immunogenic peptides.
Cell reports (2021-07-08)
Alessia Melacarne, Valentina Ferrari, Luca Tiraboschi, Michele Mishto, Juliane Liepe, Marina Aralla, Laura Marconato, Michela Lizier, Chiara Pozzi, Offer Zeira, Giuseppe Penna, Maria Rescigno
ZUSAMMENFASSUNG
Efforts to overcome resistance to immune checkpoint blockade therapy have focused on vaccination strategies using neoepitopes, although they cannot be applied on a large scale due to the "private" nature of cancer mutations. Here, we show that infection of tumor cells with Salmonella induces the opening of membrane hemichannels and the extracellular release of proteasome-generated peptides by the exacerbation of endoplasmic reticulum (ER) stress. Peptides released by cancer cells foster an antitumor response in vivo, both in mice bearing B16F10 melanomas and in dogs suffering from osteosarcoma. Mass spectrometry analysis on the supernatant of human melanoma cells revealed 12 peptides capable of priming healthy-donor CD8+ T cells that recognize and kill human melanoma cells in vitro and when xenotransplanted in vivo. Hence, we identified a class of shared tumor antigens that are generated in ER-stressed cells, such as tumor cells, that do not induce tolerance and are not presented by healthy cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatasehemmer-Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid