- Antisense mapping KOR-1: evidence for multiple kappa analgesic mechanisms.
Antisense mapping KOR-1: evidence for multiple kappa analgesic mechanisms.
Brain research (1999-05-04)
K R Pasternak, G C Rossi, A Zuckerman, G W Pasternak
PMID10224306
ZUSAMMENFASSUNG
In binding assays, both dynorphin B and alpha-neoendorphin are relatively selective for the kappa1b site, unlike U50,488H which has high affinity for both kappa1a and kappa1b sites. In vivo, U50,488H, dynorphin B and alpha-neoendorphin analgesia are reversed by the kappa1-selective antagonist, nor-binaltorphimine (norBNI). Antisense mapping the three exons of KOR-1 revealed that probes targeting all three exons blocked U50,488H analgesia, as expected. However, the selectivity profile of dynorphin B and alpha-neoendorphin analgesia towards the various antisense oligodeoxynucleotides differed markedly from U50,488H, implying a different receptor mechanism of action.