Merck
  • 3D iPSC modeling of the retinal pigment epithelium-choriocapillaris complex identifies factors involved in the pathology of macular degeneration.
3D iPSC modeling of the retinal pigment epithelium-choriocapillaris complex identifies factors involved in the pathology of macular degeneration.
Cell stem cell (2021-03-31)
Kannan V Manian, Chad A Galloway, Sonal Dalvi, Anthony A Emanuel, Jared A Mereness, Whitney Black, Lauren Winschel, Celia Soto, Yiming Li, Yuanhui Song, William DeMaria, Akhilesh Kumar, Igor Slukvin, Michael P Schwartz, William L Murphy, Bela Anand-Apte, Mina Chung, Danielle S W Benoit, Ruchira Singh
ZUSAMMENFASSUNG
The retinal pigment epithelium (RPE)-choriocapillaris (CC) complex in the eye is compromised in age-related macular degeneration (AMD) and related macular dystrophies (MDs), yet in vitro models of RPE-CC complex that enable investigation of AMD/MD pathophysiology are lacking. By incorporating iPSC-derived cells into a hydrogel-based extracellular matrix, we developed a 3D RPE-CC model that recapitulates key features of both healthy and AMD/MD eyes and provides modular control over RPE and CC layers. Using this 3D RPE-CC model, we demonstrated that both RPE- and mesenchyme-secreted factors are necessary for the formation of fenestrated CC-like vasculature. Our data show that choroidal neovascularization (CNV) and CC atrophy occur in the absence of endothelial cell dysfunction and are not necessarily secondary to drusen deposits underneath RPE cells, and CC atrophy and/or CNV can be initiated systemically by patient serum or locally by mutant RPE-secreted factors. Finally, we identify FGF2 and matrix metalloproteinases as potential therapeutic targets for AMD/MDs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Apolipoprotein E-Antikörper, serum, Chemicon®
Sigma-Aldrich
Anti-FGF-2/basic-FGF-Antikörper (neutralisierend), Klon bFM-1, clone bFM-1, Upstate®, from mouse
Sigma-Aldrich
Anti-PLVAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TIMP-3 Antibody, a.a. 170-188, clone 136-13H4, clone 136-13H4, Chemicon®, from mouse