Merck
  • Alteration of sphingosine-1-phosphate with aging induces contractile dysfunction of colonic smooth muscle cells via Ca2+ -activated K+ channel (BKCa ) upregulation.
Alteration of sphingosine-1-phosphate with aging induces contractile dysfunction of colonic smooth muscle cells via Ca2+ -activated K+ channel (BKCa ) upregulation.
Neurogastroenterology and motility (2021-01-17)
Xiaoxue Shen, Ling Zhang, Ling Jiang, Wenjie Xiong, Yurong Tang, Lin Lin, Ting Yu
ZUSAMMENFASSUNG
Age-associated changes alter calcium-activated potassium channel (BKCa ) expression of colon. Sphingolipids (SLs) are important cell membrane structural components; altered composition of SLs may affect BKCa expression. This study investigated the mechanism by which sphingosine-1-phosphate (S1P) contributes to age-associated contractile dysfunction. Fifty male Sprague Dawley rats of different ages were randomly assigned to five age-groups, namely 3, 6, 12, 18, and 24 months. BKCa expression, S1P levels, and phosphorylated myosin light chain (p-MLC) levels were tested in colonic tissues. In the absence and presence of S1P treatment, BKCa expression, p-MLC levels, and intracellular calcium mobilization were tested in vitro. BKCa small interfering RNA (siRNA) was used to investigate whether p-MLC expression and calcium mobilization were affected by BKCa in colonic smooth muscle cells (SMCs). The expressions of phosphorylated protein kinase B, c-Jun N-terminal kinases (JNKs), extracellular-regulated protein kinases, nuclear factor kappa-B (NF-κB), and protein kinase Cζ (PKCζ ) were examined to investigate the correlation between S1P and BKCa . Sphingosine-1-phosphate levels and sphingosine-1-phosphate receptor 2 (S1PR2) and BKCa expressions were upregulated and p-MLC expression was downregulated in the colonic tissues, age dependently. In the cultured SMCs, S1P treatment increased BKCa expression and reduced calcium concentration and p-MLC was observed. BKCa siRNA increased calcium concentration, and p-MLC levels significantly compared with control. We also showed that S1P upregulated BKCa through PKCζ , JNK, and NF-κB pathways. In conclusion, S1P and S1PR2 participate in age-associated contractile dysfunction via BKCa upregulation through PKCζ , JNK, and NF-κB pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin-Inhibitor aus Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Staurosporine, Streptomyces sp., Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC50 = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).
USP
Kollagenase II, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
JNK-Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
Iberiotoxin, recombinant from Mesobuthus tamulus, ≥98% (HPLC), recombinant, expressed in E. coli