Merck
  • Preparation of antioxidant enzymatic hydrolysates from honeybee-collected pollen using plant enzymes.
Preparation of antioxidant enzymatic hydrolysates from honeybee-collected pollen using plant enzymes.
Enzyme Research (2011)
MD Marinova and BP Tchorbanov