Merck
  • Non-neutralizing SARS CoV-2 directed polyclonal antibodies demonstrate cross-reactivity with the HA glycans of influenza virus.
Non-neutralizing SARS CoV-2 directed polyclonal antibodies demonstrate cross-reactivity with the HA glycans of influenza virus.
International immunopharmacology (2021-08-25)
Praveenkumar Murugavelu, Reshma Perween, Tripti Shrivastava, Vanshika Singh, Hilal Ahmad Parray, Swarandeep Singh, Adarsh Kumar Chiranjivi, Ramachandran Thiruvengadam, Savita Singh, Naveen Yadav, Kamini Jakhar, Sudipta Sonar, Shailendra Mani, Sankar Bhattacharyya, Chandresh Sharma, Preeti Vishwakarma, Ritika Khatri, Anil Kumar Panchal, Supratik Das, Shubbir Ahmed, Sweety Samal, Pallavi Kshetrapal, Shinjini Bhatnagar, Kalpana Luthra, Rajesh Kumar
ZUSAMMENFASSUNG
The spike protein of the SARS-CoV-2 virus is the foremost target for the designing of vaccines and therapeutic antibodies and also acts as a crucial antigen in the assessment of COVID-19 immune responses. The enveloped viruses; such as SARS-CoV-2, Human Immunodeficiency Virus-1 (HIV-1) and influenza, often hijack host-cell glycosylation pathways and influence pathobiology and immune selection. These glycan motifs can lead to either immune evasion or viral neutralization by the production of cross-reactive antibodies that can lead to antibody-dependent enhancement (ADE) of infection. Potential cross-protection from influenza vaccine has also been reported in COVID-19 infected individuals in several epidemiological studies recently; however, the scientific basis for these observations remains elusive. Herein, we show that the anti-SARS-CoV2 antibodies cross-reacts with the Hemagglutinin (HA) protein. This phenomenon is common to both the sera from convalescent SARS-CoV-2 donors and spike immunized mice, although these antibodies were unable to cross-neutralize, suggesting the presence of a non-neutralizing antibody response. Epitope mapping suggests that the cross-reactive antibodies are targeted towards glycan epitopes of the SARS-CoV-2 spike and HA. Overall, our findings address the cross-reactive responses, although non-neutralizing, elicited against RNA viruses and warrant further studies to investigate whether such non-neutralizing antibody responses can contribute to effector functions such as antibody-dependent cellular cytotoxicity (ADCC) or ADE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining