Merck
The antioxidative effect of fullerenes during the peroxidation of methyl linoleate in toluene.
Bioscience, biotechnology, and biochemistry (2012-01-11)
Akio Kadowaki, Satoshi Iwamoto, Ryo Yamauchi
ZUSAMMENFASSUNG
The antioxidative effect of fullerenes C(60) and C(70) was examined by measuring the inhibition of methyl linoleate (MeL) peroxidation in toluene initiated by 2,2'-azobis(2,4-dimethylvaleronitrile) (AMVN). The fullerenes retarded the formation of MeL hydroperoxides and lowered the rate of propagation. The reaction rates of fullerenes with AMVN-derived peroxyl radicals were much higher than that of MeL. These results indicate that fullerenes can act as retarders of lipid peroxidation, though their activity is low compared with that of α-tocopherol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methyllinoleat, ≥98% (GC)
Supelco
Methyllinoleat, analytical standard
Sigma-Aldrich
Buckminsterfulleren C70, 98%
Supelco
trans-9,12-Octadecadiensäuremethylester, certified reference material, 10 mg/mL in heptane
Sigma-Aldrich
Linolelaidinsäuremethylester, ≥99% (GC), liquid
Supelco
cis-9,cis-12-Octadecadiensäuremethylester, certified reference material, 10 mg/mL in heptane
Sigma-Aldrich
Buckminsterfulleren C70, sublimed, ≥99% (HPLC)
Supelco
Linolelaidinsäuremethylester, analytical standard