Merck
  • Human Corneal Epithelial Cells Internalize Aspergillus flavus Spores by Actin-Mediated Endocytosis.
Human Corneal Epithelial Cells Internalize Aspergillus flavus Spores by Actin-Mediated Endocytosis.
Infection and immunity (2021-03-24)
Divya Arunachalam, Venkatesh Prajna Namperumalsamy, Lalitha Prajna, Dharmalingam Kuppamuthu
ZUSAMMENFASSUNG
Human corneal epithelial (HCE) cells play a significant role in the innate immune response by secreting cytokines and antimicrobial peptides when they encounter fungal pathogens. But the detailed mechanism of attachment and engulfment of the fungal conidia by HCE cells is not well understood. Here, we show the phagocytosis of Aspergillus flavus conidia by RCB2280 cells and primary HCE cultures using confocal microscopy and proteomic analysis of conidium-containing phagosomes. Phalloidin staining showed actin polymerization, leading to an actin ring around engulfed conidia. Cytochalasin D inhibited the actin-mediated endocytosis of the conidia. Immunolabeling of the early endosomal markers CD71 and early endosomal antigen (EEA1) and the late endosomal markers lysosome-associated membrane protein 1 (LAMP1), Rab7, and cathepsin G showed that endosomal proteins were recruited to the site of conidia and showed maturation of the conidium-containing phagosomes. Lysotracker red DND 99 labeling showed the acidification of the phagosomes containing conidia. Phagosome-specific proteome analysis confirmed the recruitment of various phagosomal and endosomal proteins to the conidium-containing phagosomes. These results show that the ocular surface epithelium contributes actively to antifungal defense by the phagocytosis of invading fungal conidia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin aus Schweinepankreas, Proteomics Grade, BioReagent, Dimethylated