Merck
  • Purification and characterization of a novel mannitol dehydrogenase from Lactobacillus intermedius
Purification and characterization of a novel mannitol dehydrogenase from <I>Lactobacillus intermedius</I>
Biotechnology Progress (2012)
Saha, B.