Merck
Bestatin, an inhibitor of aminopeptidase B, produced by actinomycetes.
The Journal of antibiotics (1976-01-01)
H Umezawa, T Aoyagi, H Suda, M Hamada, T Takeuchi
PMID931798
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bestatin -hydrochlorid, ≥98% (HPLC)