Merck
Novel macromolecular inhibitors of human immunodeficiency virus-1 protease.
Protein engineering, design & selection : PEDS (2008-05-16)
Gabriella Miklóssy, József Tözsér, János Kádas, Rieko Ishima, John M Louis, Péter Bagossi
ZUSAMMENFASSUNG
An intracellularly expressed defective human immunodeficiency virus type-1 (HIV-1) protease (PR) monomer could function as a dominant-negative inhibitor of the enzyme that requires dimerization for activity. Based on in silico studies, two mutant PRs harboring hydrophilic mutations, capable of forming favorable inter- and intra-subunit interactions, were selected: PR(RE) containing Asp25Arg and Gly49Glu mutations, and PR(RER) containing an additional Ile50Arg mutation. The mutants were expressed and tested by PR assays, nuclear magnetic resonance (NMR) and cell culture experiments. The mutant PRs showed dose-dependent inhibition of the wild-type PR in a fluorescent microtiter plate PR assay. Furthermore, both mutants were retained by hexahistidine-tagged wild-type HIV-1 PR immobilized on nickel-chelate affinity resin. For the first time, heterodimerization between wild-type and dominant-negative mutant PRs were also demonstrated by NMR spectroscopy. (1)H-(15)N Heteronuclear Single Quantum Coherence NMR spectra showed that although PR(RE) has a high tendency to aggregate, PR(RER) exists mainly as a folded monomer at 25-35 microM concentration, but in the presence of wild-type PR in a ratio of 1:1, heterodimerization occurs with both mutants. While the recombinant virus containing the PR(RE) sequence showed only very low level of expression, expression of the viral proteins of the virus with the PR(RER) sequence was comparable with that of the wild-type. In cell culture experiments, infectivity of viral particles containing PR(RER) protein was reduced by 82%, at mutant to wild-type infective DNA ratio of 2:1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Albumin aus Rinderserum, powder, BioXtra
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, pH <5.0, powder
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, low endotoxin, pH 7, ≥98%