Merck
  • Effect of Chaiqinchengqi decoction on sarco/endoplasmic reticulum Ca2+-ATPase mRNA expression of pancreatic tissues in acute pancreatitis rats.
Effect of Chaiqinchengqi decoction on sarco/endoplasmic reticulum Ca2+-ATPase mRNA expression of pancreatic tissues in acute pancreatitis rats.
World journal of gastroenterology (2008-04-18)
Ping Xue, Li-Hui Deng, Zhao-Da Zhang, Xiao-Nan Yang, Qing Xia, Da-Kai Xiang, Lei Huang, Mei-Hua Wan
ZUSAMMENFASSUNG
To investigate the effect of Chaiqinchengqi decoction (CQCQD) on sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) mRNA expression of pancreatic tissues in acute pancreatitis (AP) rats. Thirty Sprague-Dawley (SD) rats were randomized into control group, AP group and CQCQD group (n = 3 x 10). The rats in the CQCQD group were intragastrically administered with CQCQD (10 mL/kg every 2 h) after induction of AP by intraperitoneal injection of caerulein (50 microg/kg.h x 5) within 4 h. At 6 h after the induction of AP model, pancreatic tissues were collected for the pathological observation, mRNA extraction for determination of SERCA1 and SERCA2 mRNA expression or pancreatic acinar cell isolation for measurement of fluorescence intensity (FI) of intracellular calcium ion concentration [Ca2+]i. There was no expression of pancreatic SERCA1 mRNA in the control group and the AP group. The expression of pancreatic SERCA2 mRNA in the AP group was down-regulated (expression ratio = 0.536; P = 0.001) compared with the control group, while that in the CQCQD group was up-regulated (expression ratio = 2.00; P = 0.012) compared with AP group. The FI of intracellular [Ca2+] of pancreatic acinar cells in the AP group (138.2 +/- 23.1) was higher than the C group (111.0 +/- 18.4) and the CQCQD group (118.7 +/- 15.2 ) (P < 0.05) and the pancreatic pathological score in the CQCQD group was lower than that in the AP group (5.7 +/- 1.9 vs 9.2 +/- 2.7, P < 0.05). CQCQD can up-regulate the expression of SERCA2 mRNA of pancreatic tissues, reduce intracellular calcium overload and relieve pancreatic tissue lesions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caerulein, ≥95% (HPLC)