Merck
  • Protective effects of galangin against H2O2/UVB-induced dermal fibroblast collagen degradation via hsa-microRNA-4535-mediated TGFβ/Smad signaling.
Protective effects of galangin against H2O2/UVB-induced dermal fibroblast collagen degradation via hsa-microRNA-4535-mediated TGFβ/Smad signaling.
Aging (2021-12-11)
Jian-Jr Lee, Shang-Chuan Ng, Yean-Tin Ni, Jian-Sheng Liu, Chih-Jung Chen, Viswanadha Vijaya Padma, Chih-Yang Huang, Wei-Wen Kuo
ZUSAMMENFASSUNG
This study aimed to investigate the mechanism underlying the protective effects of galangin against H2O2/UVB-induced damage using in vitro and in vivo models of photodamage. Moreover, we identified the involvement of miRNA regulation in this process. The H2O2/UVB-treated HS68 human dermal fibroblasts and UVB-induced C57BL/6J nude mice were used as in vitro and in vivo models of photodamage. The results showed that galangin treatment alleviated H2O2/UVB-induced reduction in cell viability, TGFβ/Smad signaling impairment, and dermal aging. Based on the results of microRNA array analyses and database searches, hsa-miR-4535 was identified as a potential candidate miRNA that targets Smad4. In vitro, galangin treatment activated Smad2/3/4 complex and inhibited hsa-miR-4535 expression in H2O2/UVB-exposed cells. In vivo, topical application of low (12 mg/kg) and high doses (24 mg/kg) of galangin to the dorsal skin of C57BL/6J nude mice significantly alleviated UVB-induced skin photodamage by promoting TGFβ/Smad collagen synthesis signaling, reducing epidermal hyperplasia, wrinkle formation, and skin senescence, as well as inhibiting hsa-miR-4535 expression. Taken together, our findings indicate a link between hsa-miR-4535 and TGFβ/Smad collagen synthesis signaling and suggest these factors to be involved in the photo-protective mechanism of galangin in dermal fibroblasts against H2O2/UVB-induced aging. The evidence indicated that galangin with anti-aging properties can be considered as a supplement in skin care products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Mitochondria Staining Kit, 1 kit sufficient for 40 tests (of 5 mL cell suspensions), 1 kit sufficient for 200 tests (of 1 mL cell suspensions)
Sigma-Aldrich
Galangin, autophagy inducing flavonoid