- Valosin: isolation and characterization of a novel peptide from porcine intestine.
Valosin: isolation and characterization of a novel peptide from porcine intestine.
FEBS letters (1985-10-28)
W E Schmidt, V Mutt, M Carlquist, H Kratzin, J M Conlon, W Creutzfeldt
PMID4054310
ZUSAMMENFASSUNG
The isolation and primary structure of a novel gastrointestinal peptide, designated valosin, is described. The peptide was purified from porcine upper gut extracts using an HPLC and N-terminal sequence screening strategy which depends on chromatographic and structural characteristics as isolation criterion. The amino acid sequence of this peptide consists of 25 amino acid residues: