Merck
  • Naturally occurring novel promoters around pyruvate branch-point for recombinant protein production in Pichia pastoris (Komagataella phaffii): pyruvate decarboxylase-and pyruvate kinase-promoters
Naturally occurring novel promoters around pyruvate branch-point for recombinant protein production in Pichia pastoris (Komagataella phaffii): pyruvate decarboxylase-and pyruvate kinase-promoters
Biochemical Engineering Journal (2018)
Massahi A and Calik P