Merck
  • Intensified training in adolescent female athletes: a crossover study of Greek yogurt effects on indices of recovery.
Intensified training in adolescent female athletes: a crossover study of Greek yogurt effects on indices of recovery.
Journal of the International Society of Sports Nutrition (2022-05-24)
Brandon J McKinlay, Phillip J Wallace, Shai Olansky, Stacey Woods, Nigel Kurgan, Brian D Roy, Andrea R Josse, Bareket Falk, Panagiota Klentrou
ZUSAMMENFASSUNG
During a period of intensified exercise (e.g. training/identification camps), often undertaken by competitive youth athletes, the maintenance of muscle function and peak performance can become challenging due to an accumulation of fatigue. The provision of post-exercise dairy protein in adults has been previously shown to accelerate recovery; however, its efficacy in youth athletes is currently unknown. Therefore, the purpose of this study was to examine the effects of increased dairy protein consumption with plain Greek yogurt (GY) on performance and recovery indices during an intensified soccer training camp in adolescent female soccer players. Thirteen players (14.3 ± 1.3 years) participated in a randomized, double blinded, crossover design study where they received 3 servings/day of either GY (~115 kcal, 17 g protein, ~11.5 g carbohydrates) or an isoenergetic carbohydrate control (CHO, ~115 kcal, 0.04 g protein, ~28.6 g carbohydrates) during two 5-day soccer-specific training camps. Performance was assessed before and after each training camp. Fasted, morning, creatine kinase (CK), insulin-like growth factor-1 (IGF-1), C-reactive protein (CRP), interleukin 6 (IL6), interleukin 10 (IL10) and tumor necrosis factor-α (TNFα) were measured in plasma pre- and post-training. Training led to decrements in counter-movement jump (p = 0.01), broad jump (p = 0.04) and aerobic capacity (p = 0.006), with no effect of GY. A significant increase in anti-inflammatory cytokine IL10 was observed from pre- to post-training in GY (+26% [p = 0.008]) but not in CHO (p = 0.89). CRP and CK increased (+65% [p = 0.005] and +119% [p ≤ 0.001], respectively), while IGF-1 decreased (-34% [p ≤ 0.001]) from pre- to post-training with no difference between conditions. These results demonstrate that consumption of GY did not offer any added recovery benefit with respect to measures of performance and in the attenuation of exercise-induced muscle damage above that achieved with energy-matched carbohydrate in this group of young female soccer players. However, regular consumption of GY may assist with the acute anti-inflammatory response during periods of intensified training in adolescent athletes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel, Configurable 96-well Human High Sensitivity T Cell 21-Plex Panel