Merck
Exposure Route Influences Disease Severity in the COVID-19 Cynomolgus Macaque Model.
Viruses (2022-05-29)
Sandra L Bixler, Christopher P Stefan, Alexandra N Jay, Franco D Rossi, Keersten M Ricks, Charles J Shoemaker, Alicia M Moreau, Xiankun Zeng, Jay W Hooper, David N Dyer, Ondraya M Frick, Jeffrey W Koehler, Brian J Kearney, Nina DiPinto, Jun Liu, Samantha D Tostenson, Tamara L Clements, Jeffrey M Smith, Joshua A Johnson, Kerry L Berrier, Heather L Esham, Korey L Delp, Susan R Coyne, Holly A Bloomfield, Paul A Kuehnert, Kristen Akers, Kathleen M Gibson, Timothy D Minogue, Aysegul Nalca, Margaret L M Pitt
ZUSAMMENFASSUNG
The emergence of SARS-CoV-2 and the subsequent pandemic has highlighted the need for animal models that faithfully replicate the salient features of COVID-19 disease in humans. These models are necessary for the rapid selection, testing, and evaluation of potential medical countermeasures. Here, we performed a direct comparison of two distinct routes of SARS-CoV-2 exposure-combined intratracheal/intranasal and small particle aerosol-in two nonhuman primate species, rhesus and cynomolgus macaques. While all four experimental groups displayed very few outward clinical signs, evidence of mild to moderate respiratory disease was present on radiographs and at necropsy. Cynomolgus macaques exposed via the aerosol route also developed the most consistent fever responses and had the most severe respiratory disease and pathology. This study demonstrates that while all four models produced suitable representations of mild COVID-like illness, aerosol exposure of cynomolgus macaques to SARS-CoV-2 produced the most severe disease, which may provide additional clinical endpoints for evaluating therapeutics and vaccines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-MxA, clone M143 (CL143), clone CL143, from mouse
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−R-Phycoerythrin antibody produced in goat, affinity isolated antibody, buffered aqueous solution