Merck
Bacteria induce skin regeneration via IL-1β signaling.
Cell host & microbe (2021-04-03)
Gaofeng Wang, Evan Sweren, Haiyun Liu, Eric Wier, Martin P Alphonse, Ruosi Chen, Nasif Islam, Ang Li, Yingchao Xue, Junjie Chen, Seungman Park, Yun Chen, Sam Lee, Yu Wang, Saifeng Wang, Nate K Archer, William Andrews, Maureen A Kane, Erika Dare, Sashank K Reddy, Zhiqi Hu, Elizabeth A Grice, Lloyd S Miller, Luis A Garza
ZUSAMMENFASSUNG
Environmental factors that enhance regeneration are largely unknown. The immune system and microbiome are attributed roles in repairing and regenerating structure but their precise interplay is unclear. Here, we assessed the function of skin bacteria in wound healing and wound-induced hair follicle neogenesis (WIHN), a rare adult organogenesis model. WIHN levels and stem cell markers correlate with bacterial counts, being lowest in germ-free (GF), intermediate in conventional specific pathogen-free (SPF), and highest in wild-type mice, even those infected with pathogenic Staphylococcus aureus. Reducing skin microbiota via cage changes or topical antibiotics decreased WIHN. Inflammatory cytokine IL-1β and keratinocyte-dependent IL-1R-MyD88 signaling are necessary and sufficient for bacteria to promote regeneration. Finally, in a small trial, a topical broad-spectrum antibiotic also slowed skin wound healing in adult volunteers. These results demonstrate a role for IL-1β to control morphogenesis and support the need to reconsider routine applications of topical prophylactic antibiotics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Liberase TL, Forschungsqualität, lyophilized, suitable for tissue processing, optimum pH 7.4
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Anti-KRT15 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution