Merck
  • Cardiac glycoside-mediated turnover of Na, K-ATPases as a rational approach to reducing cell surface levels of the cellular prion protein.
Cardiac glycoside-mediated turnover of Na, K-ATPases as a rational approach to reducing cell surface levels of the cellular prion protein.
PloS one (2022-07-02)
Mohadeseh Mehrabian, Xinzhu Wang, Shehab Eid, Bei Qi Yan, Mark Grinberg, Murdock Siegner, Christopher Sackmann, Muhammad Sulman, Wenda Zhao, Declan Williams, Gerold Schmitt-Ulms
ZUSAMMENFASSUNG
It is widely anticipated that a reduction of brain levels of the cellular prion protein (PrPC) can prolong survival in a group of neurodegenerative diseases known as prion diseases. To date, efforts to decrease steady-state PrPC levels by targeting this protein directly with small molecule drug-like compounds have largely been unsuccessful. Recently, we reported Na,K-ATPases to reside in immediate proximity to PrPC in the brain, unlocking an opportunity for an indirect PrPC targeting approach that capitalizes on the availability of potent cardiac glycosides (CGs). Here, we report that exposure of human co-cultures of neurons and astrocytes to non-toxic nanomolar levels of CGs causes profound reductions in PrPC levels. The mechanism of action underpinning this outcome relies primarily on a subset of CGs engaging the ATP1A1 isoform, one of three α subunits of Na,K-ATPases expressed in brain cells. Upon CG docking to ATP1A1, the ligand receptor complex, and PrPC along with it, is internalized by the cell. Subsequently, PrPC is channeled to the lysosomal compartment where it is digested in a manner that can be rescued by silencing the cysteine protease cathepsin B. These data signify that the repurposing of CGs may be beneficial for the treatment of prion disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
MG-132, gebrauchsfertige Lösung, ≥90% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial
Sigma-Aldrich
Bafilomycin A1 Fertiglösung, 0.16 mM in DMSO, from Streptomyces griseus
Sigma-Aldrich
Ouabain Octahydrat, ≥95% (HPLC), powder
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Anti-Prion-Proteinantikörper, AS 109–112, Klon 3F4, clone 3F4, Chemicon®, from mouse
Sigma-Aldrich
Kalpain-Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)