Merck
Role of CXCR4 and SDF-1 in mammary tumor metastasis in the cat.
The Journal of veterinary medical science (2003-11-06)
Toshina Oonuma, Masami Morimatsu, Takayuki Nakagawa, Rina Uyama, Nobuo Sasaki, Munekazu Nakaichi, Hirokazu Tamamura, Nobutaka Fujii, Shizu Hashimoto, Hozumi Yamamura, Bunei Syuto
ZUSAMMENFASSUNG
It has recently been suggested that the chemokine receptor CXCR4 and its ligand SDF-1 (CXCL12) promote metastasis of various cancers in humans. Since feline mammary tumors also metastasize to distant organs frequently, we used real-time quantitative PCR to examine the expression of feline CXCR4 (fCXCR4) in ten feline mammary tumor cell lines and seven feline mammary tumor tissues, and also the expression of feline SDF-1 (fSDF-1) in various organs. Cell lines derived from metastatic regions expressed more fCXCR4 than those derived from primary tumors. Mammary tumor tissues overexpressed more fCXCR4 than normal mammary tissues. Organs with high levels of fSDF-1 expression represent common sites of metastasis. Migration assays using the feline mammary tumor cell line NAC were also performed to test the activity of TN14003 and TC14012, antagonists of human CXCR4, to antagonize fCXCR4 expressed on NAC cells. TN14003 and TC14012 inhibited migration of NAC cells. We conclude that fCXCR4 may be a therapeutic target for feline mammary tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TC14012 Trifluoroacetate, ≥95% (HPLC)