Merck
Structural and Molecular Basis for Katanin-Mediated Severing of Glutamylated Microtubules.
Cell reports (2019-01-31)
Sang Chul Shin, Sun-Kyoung Im, Eun-Hae Jang, Kyeong Sik Jin, Eun-Mi Hur, Eunice EunKyeong Kim
ZUSAMMENFASSUNG
Katanin was the first microtubule (MT)-severing enzyme discovered, but how katanin executes MT severing remains poorly understood. Here, we report X-ray crystal structures of the apo and ATPγS-bound states of the catalytic AAA domain of human katanin p60 at 3.0 and 2.9 Å resolution, respectively. Comparison of the two structures reveals conformational changes induced by ATP binding and how such changes ensure hexamer stability. Moreover, we uncover structural details of pore loops (PLs) and show that Arg283, a residue unique to katanin among MT-severing enzymes, protrudes from PL1 and lines the entry of the catalytic pore. Functional studies suggest that PL1 and Arg283 play essential roles in the recognition and remodeling of the glutamylated, C-terminal tubulin tail and regulation of axon growth. In addition, domain-swapping experiments in katanin and spastin suggest that the non-homologous N-terminal region, which contains the MT-interacting and trafficking domain and a linker, confers specificity to the severing process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatasehemmer-Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatasehemmer-Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
5-Fluor-2′-Desoxyuridin, thymidylate synthase inhibitor
Sigma-Aldrich
Rosetta (DE3)-Kompetente Zellen – Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sigma-Aldrich
Anti-β-Tubulin III-Antikörper (neuronal), monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tubulin-Antikörper, Klon YL1/2, clone YL1/2, Chemicon®, from rat