Merck
Unlocking SARS-CoV-2 detection in low- and middle-income countries.
Cell reports methods (2021-10-27)
Roberto Alcántara, Katherin Peñaranda, Gabriel Mendoza-Rojas, Jose A Nakamoto, Johanna Martins-Luna, Juana Del Valle-Mendoza, Vanessa Adaui, Pohl Milón
ZUSAMMENFASSUNG
Low- and middle-income countries (LMICs) are significantly affected by SARS-CoV-2, partially due to their limited capacity for local production and implementation of molecular testing. Here, we provide detailed methods and validation of a molecular toolkit that can be readily produced and deployed using laboratory equipment available in LMICs. Our results show that lab-scale production of enzymes and nucleic acids can supply over 50,000 tests per production batch. The optimized one-step RT-PCR coupled to CRISPR-Cas12a-mediated detection showed a limit of detection of 102 ge/μL in a turnaround time of 2 h. The clinical validation indicated an overall sensitivity of 80%-88%, while for middle and high viral load samples (Cq ≤ 31) the sensitivity was 92%-100%. The specificity was 96%-100% regardless of viral load. Furthermore, we show that the toolkit can be used with the mobile laboratory Bento Lab, potentially enabling LMICs to implement detection services in unattended remote regions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
EDTA, Dinatriumsalz, Dihydrat, molekularbiologische Qualität