Merck
Neutrophils direct preexisting matrix to initiate repair in damaged tissues.
Nature immunology (2022-04-01)
Adrian Fischer, Juliane Wannemacher, Simon Christ, Tim Koopmans, Safwen Kadri, Jiakuan Zhao, Mahesh Gouda, Haifeng Ye, Martin Mück-Häusl, Peter W Krenn, Hans-Günther Machens, Reinhard Fässler, Philipp-Alexander Neumann, Stefanie M Hauck, Yuval Rinkevich
ZUSAMMENFASSUNG
Internal organs heal injuries with new connective tissue, but the cellular and molecular events of this process remain obscure. By tagging extracellular matrix around the mesothelium lining in mouse peritoneum, liver and cecum, here we show that preexisting matrix was transferred across organs into wounds in various injury models. Using proteomics, genetic lineage-tracing and selective injury in juxtaposed organs, we found that the tissue of origin for the transferred matrix likely dictated the scarring or regeneration of the healing tissue. Single-cell RNA sequencing and genetic and chemical screens indicated that the preexisting matrix was transferred by neutrophils dependent on the HSF-integrin AM/B2-kindlin3 cascade. Pharmacologic inhibition of this axis prevented matrix transfer and the formation of peritoneal adhesions. Matrix transfer was thus an early event of wound repair and provides a therapeutic window to dampen scaring across a range of conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gelatine -Lösung, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-KINDLIN3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution