Merck
  • ALKBH3 is dispensable in maintaining hematopoietic stem cells but forced ALKBH3 rectified the differentiation skewing of aged hematopoietic stem cells.
ALKBH3 is dispensable in maintaining hematopoietic stem cells but forced ALKBH3 rectified the differentiation skewing of aged hematopoietic stem cells.
Blood science (Baltimore, Md.) (2020-10-23)
Hanqing He, Yuqian Wang, Jianwei Wang
ZUSAMMENFASSUNG
ALKBH3, a demethylase responsible for demethylating N1-methyladenosine (m1A) in mRNA and N1-methyldeoxyadenosine in single-stranded DNA, plays an important role in DNA repair and cancer cell proliferation. However, its function in hematopoietic stem cells (HSCs) is unknown. In this study, we generated Alkbh3 knockout mice and observed that the deletion of Alkbh3 does not impair the reconstitution capacity of HSCs in both primary and secondary transplantation. Aged hematopoietic stem and progenitor cells exhibit increased expression of ALKBH3. Forced ALKBH3 rescued the differentiation skewing without affecting the reconstitution capacity of aged HSCs. In brief, our study for the first time investigated the functional role of ALKBH3 in hematopoietic system, and observed that ALKBH3 is dispensable for HSCs maintenance and differentiation, but overexpression of ALKBH3 rectified the differentiation skewing of aged HSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence