Merck
  • Distinct diagnostic trajectories in NBAS-associated acute liver failure highlights the need for timely functional studies.
Distinct diagnostic trajectories in NBAS-associated acute liver failure highlights the need for timely functional studies.
JIMD reports (2022-04-19)
Lauren S Akesson, Rocio Rius, Natasha J Brown, Jeremy Rosenbaum, Sarah Donoghue, Michael Stormon, Charmaine Chai, Esmeralda Bordador, Yiran Guo, Hakon Hakonarson, Alison G Compton, David R Thorburn, Sumudu Amarasekera, Justine Marum, Alisha Monaco, Crystle Lee, Belinda Chong, Sebastian Lunke, Zornitza Stark, John Christodoulou
ZUSAMMENFASSUNG
Variants of uncertain significance (VUS) are commonly found following genomic sequencing, particularly in ethnically diverse populations that are underrepresented in large population databases. Functional characterization of VUS may assist in variant reclassification, however these studies are not readily available and often rely on research funding and good will. We present four individuals from three families at different stages of their diagnostic trajectory with recurrent acute liver failure (RALF) and biallelic NBAS variants, confirmed by either trio analysis or cDNA studies. Functional characterization was undertaken, measuring NBAS and p31 levels by Western blotting, demonstrating reduced NBAS levels in two of three families, and reduced p31 levels in all three families. These results provided functional characterization of the molecular impact of a missense VUS, allowing reclassification of the variant and molecular confirmation of NBAS-associated RALF. Importantly, p31 was decreased in all individuals, including an individual with two missense variants where NBAS protein levels were preserved. These results highlight the importance of access to timely functional studies after identification of putative variants, and the importance of considering a range of assays to validate variants whose pathogenicity is uncertain. We suggest that funding models for genomic sequencing should consider incorporating capabilities for adjunct RNA, protein, biochemical, and other specialized tests to increase the diagnostic yield which will lead to improved medical care, increased equity, and access to molecular diagnoses for all patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GAPDH in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-USE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution