Merck
  • Creatine-mediated crosstalk between adipocytes and cancer cells regulates obesity-driven breast cancer.
Creatine-mediated crosstalk between adipocytes and cancer cells regulates obesity-driven breast cancer.
Cell metabolism (2021-02-18)
Olivia A Maguire, Sarah E Ackerman, Sarah K Szwed, Aarthi V Maganti, François Marchildon, Xiaojing Huang, Daniel J Kramer, Adriana Rosas-Villegas, Rebecca G Gelfer, Lauren E Turner, Victor Ceballos, Asal Hejazi, Bozena Samborska, Janane F Rahbani, Christien B Dykstra, Matthew G Annis, Ji-Dung Luo, Thomas S Carroll, Caroline S Jiang, Andrew J Dannenberg, Peter M Siegel, Sarah A Tersey, Raghavendra G Mirmira, Lawrence Kazak, Paul Cohen
ZUSAMMENFASSUNG
Obesity is a major risk factor for adverse outcomes in breast cancer; however, the underlying molecular mechanisms have not been elucidated. To investigate the role of crosstalk between mammary adipocytes and neoplastic cells in the tumor microenvironment (TME), we performed transcriptomic analysis of cancer cells and adjacent adipose tissue in a murine model of obesity-accelerated breast cancer and identified glycine amidinotransferase (Gatm) in adipocytes and Acsbg1 in cancer cells as required for obesity-driven tumor progression. Gatm is the rate-limiting enzyme in creatine biosynthesis, and deletion in adipocytes attenuated obesity-driven tumor growth. Similarly, genetic inhibition of creatine import into cancer cells reduced tumor growth in obesity. In parallel, breast cancer cells in obese animals upregulated the fatty acyl-CoA synthetase Acsbg1 to promote creatine-dependent tumor progression. These findings reveal key nodes in the crosstalk between adipocytes and cancer cells in the TME necessary for obesity-driven breast cancer progression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial