Merck
Single-nuclei isoform RNA sequencing unlocks barcoded exon connectivity in frozen brain tissue.
Nature biotechnology (2022-03-09)
Simon A Hardwick, Wen Hu, Anoushka Joglekar, Li Fan, Paul G Collier, Careen Foord, Jennifer Balacco, Samantha Lanjewar, Maureen McGuirk Sampson, Frank Koopmans, Andrey D Prjibelski, Alla Mikheenko, Natan Belchikov, Julien Jarroux, Anne Bergstrom Lucas, Miklós Palkovits, Wenjie Luo, Teresa A Milner, Lishomwa C Ndhlovu, August B Smit, John Q Trojanowski, Virginia M Y Lee, Olivier Fedrigo, Steven A Sloan, Dóra Tombácz, M Elizabeth Ross, Erich Jarvis, Zsolt Boldogkői, Li Gan, Hagen U Tilgner
ZUSAMMENFASSUNG
Single-nuclei RNA sequencing characterizes cell types at the gene level. However, compared to single-cell approaches, many single-nuclei cDNAs are purely intronic, lack barcodes and hinder the study of isoforms. Here we present single-nuclei isoform RNA sequencing (SnISOr-Seq). Using microfluidics, PCR-based artifact removal, target enrichment and long-read sequencing, SnISOr-Seq increased barcoded, exon-spanning long reads 7.5-fold compared to naive long-read single-nuclei sequencing. We applied SnISOr-Seq to adult human frontal cortex and found that exons associated with autism exhibit coordinated and highly cell-type-specific inclusion. We found two distinct combination patterns: those distinguishing neural cell types, enriched in TSS-exon, exon-polyadenylation-site and non-adjacent exon pairs, and those with multiple configurations within one cell type, enriched in adjacent exon pairs. Finally, we observed that human-specific exons are almost as tightly coordinated as conserved exons, implying that coordination can be rapidly established during evolution. SnISOr-Seq enables cell-type-specific long-read isoform analysis in human brain and in any frozen or hard-to-dissociate sample.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining