Merck
Tetraspanin CD82 restrains phagocyte migration but supports macrophage activation.
iScience (2022-06-28)
Erin N S McGowan, Osanna Wong, Eleanor Jones, Julie Nguyen, Janet Wee, Maria C Demaria, Devy Deliyanti, Chad J Johnson, Michael J Hickey, Malcolm J McConville, Jennifer L Wilkinson-Berka, Mark D Wright, Katrina J Binger
ZUSAMMENFASSUNG
Phagocytes migrate into tissues to combat infection and maintain tissue homeostasis. As dysregulated phagocyte migration and function can lead to inflammation or susceptibility to infection, identifying molecules that control these processes is critical. Here, we show that the tetraspanin CD82 restrains the migration of neutrophils and macrophages into tissues. Cd82 -/- phagocytes exhibited excessive migration during in vivo models of peritoneal inflammation, superfusion of CXCL1, retinopathy of prematurity, and infection with the protozoan parasite L. mexicana. However, with the latter, while Cd82 -/- macrophages infiltrated infection sites at higher proportions, cutaneous L. mexicana lesions were larger and persisted, indicating a failure to control infection. Analyses of in vitro bone-marrow-derived macrophages showed CD82 deficiency altered cellular morphology, and impaired gene expression and metabolism in response to anti-inflammatory activation. Altogether, this work reveals an important role for CD82 in restraining phagocyte infiltration and mediating their differentiation in response to stimulatory cues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Pferdeserum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Lectin ausBandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder