Merck
  • An antibody-based proximity labeling protocol to identify biotinylated interactors of SARS-CoV-2.
An antibody-based proximity labeling protocol to identify biotinylated interactors of SARS-CoV-2.
STAR protocols (2022-05-26)
Limin Shang, Yuehui Zhang, Yuchen Liu, Chaozhi Jin, Yanan Zhao, Jing Zhang, Pei-Hui Wang, Jian Wang
ZUSAMMENFASSUNG
Elucidating the molecular interactions between virus and host is fundamental to understanding the mechanism of viral pathogenesis. Here, we present a protocol to screen SARS-CoV-2 protein interactors using an antibody-based TurboID proximity labeling approach. This technique directly identifies biotinylated peptides labeled by the TurboID-tagged viral proteins. We describe the steps to prepare biotinylated peptide samples for mass spectrometry analysis and a stringent workflow to identify biotinylated high-confidence interactors of the virus by filtering out non-specific co-purified proteins. For complete details on the use and execution of this protocol, please refer to Zhang et al. (2022).