Merck
  • A simple HPLC bioanalytical method for the determination of doxorubicin hydrochloride in rat plasma: application to pharmacokinetic studies
A simple HPLC bioanalytical method for the determination of doxorubicin hydrochloride in rat plasma: application to pharmacokinetic studies
Tropical Journal of Pharmaceutical Research (2014)
Dharmalingam SR, et al.