Merck
  • A Protocol for the Acquisition of Comprehensive Proteomics Data from Single Cases Using Formalin-Fixed Paraffin Embedded Sections.
A Protocol for the Acquisition of Comprehensive Proteomics Data from Single Cases Using Formalin-Fixed Paraffin Embedded Sections.
Methods and protocols (2022-07-28)
Mitchell Acland, Parul Mittal, Georgia Arentz, Fergus Whitehead, Peter Hoffmann, Manuela Klingler-Hoffmann, Martin K Oehler
ZUSAMMENFASSUNG
The molecular analysis of small or rare patient tissue samples is challenging and often limited by available technologies and resources, such as reliable antibodies against a protein of interest. Although targeted approaches provide some insight, here, we describe the workflow of two complementary mass spectrometry approaches, which provide a more comprehensive and non-biased analysis of the molecular features of the tissue of interest. Matrix-assisted laser desorption/ionization (MALDI) mass spectrometry imaging (MSI) generates spatial intensity maps of molecular features, which can be easily correlated with histology. Additionally, liquid chromatography tandem mass spectrometry (LC-MS/MS) can identify and quantify proteins of interest from a consecutive section of the same tissue. Here, we present data from concurrent precancerous lesions from the endometrium and fallopian tube of a single patient. Using this complementary approach, we monitored the abundance of hundreds of proteins within the precancerous and neighboring healthy regions. The method described here represents a useful tool to maximize the number of molecular data acquired from small sample sizes or even from a single case. Our initial data are indicative of a migratory phenotype in these lesions and warrant further research into their malignant capabilities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[Glu1]-Fibrinopeptid B, ≥90% (HPLC)
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 1-24 human, rat, ≥97% (HPLC)